To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:ATF6
- Clonality:Monoclonal
- Clone:70B1413.1
- Conjugation:DyLight® 680
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Fish,Hamster,Rabbit,Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:22926
- Antigen Synonyms:cAMP-dependent transcription factor ATF-6 alpha|ATF6-alpha|cyclic AMP-dependent transcription factor ATF-6 alpha|atf6 a|ATF6A|activating transcription factor 6|Activating transcription factor 6 alpha
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:This monoclonal antibody was made against a partial protein containing amino acids 1-273 of human ATF6.
- Purification:Protein G purified
- Cat. No.:103405-308
Specifications
About this item
The ATF6 Antibody (70B1413.1) [DyLight 680] from Novus Biologicals is a mouse monoclonal antibody to ATF6. This antibody reacts with human, mouse, rat, fish, hamster, rabbit. The ATF6 Antibody (70B1413.1) [DyLight 680] has been validated for the following applications: Western Blot, Flow Cytometry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Type: Primary
Antigen: ATF6
Clonality: Monoclonal
Clone: 70B1413.1
Conjugation: DyLight 680
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat, Fish, Hamster, Rabbit