To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Activating enhancer-binding protein 2-alpha
- Antigen Symbol:AP2 alpha
- Clonality:Monoclonal
- Clone:A6/2/2
- Conjugation:DyLight® 755
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:7020
- Antigen Synonyms:transcription factor AP-2-alpha|AP-2 transcription factor|BOFS|AP2-alpha|transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha)|AP2TFAP-2alpha|Activator protein 2|Activating enhancer-binding protein 2-alpha|TFAP2|AP-2transcription factor AP-2 alpha (activating enhancer-binding protein 2 alpha)|FLJ51761
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Peptide to C-Terminal of AP-2 Alpha coupled to KLH. PNSHTDNNAKSSDKEEKHRK
- Purification:Protein A purified
- Cat. No.:103413-476
Specifications
About this item
The AP2 alpha Antibody (A6 / 2 / 2) [DyLight 755] from Novus Biologicals is a mouse monoclonal antibody to AP2 alpha. This antibody reacts with human. The AP2 alpha Antibody (A6 / 2 / 2) [DyLight 755] has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: AP2 alpha
Clonality: Monoclonal
Clone: A6/2/2
Conjugation: DyLight 755
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human