To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Alkaline Phosphatase
- Antigen Symbol:ALPP
- Clonality:Monoclonal
- Clone:H17E2
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 μl
- Environmentally Preferable:
- Gene ID:250
- Antigen Synonyms:Regan isozyme|PALP|Placental alkaline phosphatase 1|EC 3.1.3.1|alkaline phosphatase|ALP|alkaline phosphomonoesterase|placental|FLJ61142|PLAP|PLAP-1|glycerophosphatase|placental type|Alkaline phosphatase Regan isozyme|placental (Regan isozyme)
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Placental membrane (Human).
- Purification:Protein G purified
- Cat. No.:103412-826
Specifications
About this item
The Alkaline Phosphatase / ALPP Antibody (H17E2) [PerCP] from Novus Biologicals is a mouse monoclonal antibody to Alkaline Phosphatase / ALPP. This antibody reacts with human. The Alkaline Phosphatase / ALPP Antibody (H17E2) [PerCP] has been validated for the following applications: Flow Cytometry, ELISA, Immunohistochemistry-Frozen.
Type: Primary
Antigen: ALPP
Clonality: Monoclonal
Clone: H17E2
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human