To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:AG-2
- Clonality:Monoclonal
- Clone:1000
- Conjugation:DyLight® 755
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Primates,Horse,Human,Bovine,Dog,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:10551
- Antigen Synonyms:AG2member 17|secreted cement gland homolog|hAG-2|AG-2|anterior gradient 2 homolog (Xenopus laevis)|anterior gradient homolog 2 (Xenopus laevis)|HPC8|GOB-4
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:A portion of amino acids 50-100 of human AGR2 was used as the immunogen for the antibody.
- Purification:Protein G purified
- Cat. No.:103409-866
Specifications
About this item
The AG-2 Antibody (10E2) [DyLight 755] from Novus Biologicals is a mouse monoclonal antibody to AG-2. This antibody reacts with human, mouse, bovine, canine, equine, other, primate. The AG-2 Antibody (10E2) [DyLight 755] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: AG-2
Clonality: Monoclonal
Clone: 1000
Conjugation: DyLight 755
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Bovine, Canine, Equine, Other, Primate