To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:ABCB5
- Clonality:Monoclonal
- Clone:5H3C6
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:340273
- Antigen Synonyms:ABCB5alpha|P-glycoprotein ABCB5|ATP-binding cassette sub-family B member 5|EST422562|ATP-binding cassette protein|ABCB5beta|EC 3.6.3.44|EC 3.6.3|ABCB5 P-gp|member 5|ATP-binding cassette|sub-family B (MDR/TAP)
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Purified recombinant fragment of human ABCB5 (AA: 481-674) expressed in E. Coli.
- Purification:Protein A or G purified
- Cat. No.:103408-030
The ABCB5 Antibody (5H3C6) from Novus Biologicals is a mouse monoclonal antibody to ABCB5. This antibody reacts with human. The ABCB5 Antibody (5H3C6) has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.
CyTOF-ready
Type: Primary
Antigen: ABCB5
Clonality: Monoclonal
Clone: 5H3C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human