To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:CYP1A2
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg/vial
- Environmentally Preferable:
- Form:Lyophilized
- Gene ID:P05177
- Molecular Weight:58294
- Storage Temperature:At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
- Concentration:Add 0.2ml of distilled water will yield a concentration of 500 μg/ml.
- Shipping Temperature:4°C
- Immunogen:E.coli-derived human CYP1A2 recombinant protein (Position: H185-D320). Human CYP1A2 shares 69.9% and 72.1% amino acid (aa) sequence identity with mouse and rat CYP1A2, respectively.
- Purification:Immunogen affinity purified.
- Cat. No.:76173-956
Rabbit IgG polyclonal antibody for Cytochrome P450 1A2(CYP1A2) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Type: Primary
Antigen: CYP1A2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human; Mouse; Rat