Order Entry
United States
ContactUsLinkComponent
Anti-Livin Polyclonal Antibody
Anti-Livin Polyclonal Antibody
Catalog # 76174-952
undefined
Anti-Livin Polyclonal Antibody
Catalog # 76174-952
Supplier Number:  RP1081
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    Livin
  • Clonality:
    Polyclonal
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Environmentally Preferable:
  • Cat. No.:
    76174-952

Specifications

About this item

Polyclonal antibody for Livin/BIRC7 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: ELISA. Reactive species: Human. Livin/BIRC7 information: Molecular Weight: 32798 MW; Subcellular Localization: Nucleus . Cytoplasm . Golgi apparatus . Nuclear, and in a filamentous pattern throughout the cytoplasm. Full-length livin is detected exclusively in the cytoplasm, whereas the truncated form (tLivin) is found in the peri-nuclear region with marked localization to the Golgi apparatus; the accumulation of tLivin in the nucleus shows positive correlation with the increase in apoptosis; Tissue Specificity: Isoform 1 and isoform 2 are expressed at very low levels or not detectable in most adult tissues. Detected in adult heart, placenta, lung, lymph node, spleen and ovary, and in several carcinoma cell lines. Isoform 2 is detected in fetal kidney, heart and spleen, and at lower levels in adult brain, skeletal muscle and peripheral blood leukocytes.

Type: Primary
Antigen: Livin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human