To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Biotin
- Source:
- Species:
- Size:1 mg
- Environmentally Preferable:
- Storage Conditions:–20 °C
- Protein Synonyms:RGD Peptide LC-biotin|GRGDS Peptide
- Protein/Peptide Name:GRGDS, LC-biotin labeled
- Purity:95%
- Molecular Weight:830 Da
- Sequence:Biotin-LC-GRGDS
- Cat. No.:103007-304
- Supplier no.:AS-62347
Specifications
About this item
The GRGDS peptide contains the amino acid sequence Arg-Gly-Asp (RGD), which has been implicated as a recognition site in interactions between extracellular matrix (ECM) molecules and cell membrane receptors. RGD-containing synthetic peptides are known to inhibit attachment of endothelial cells to substrates. GRGDS peptide inhibits angiogenesis in serum-free collagen gel culture. This synthetic peptide mimics the cellular binding site of many adhesive proteins in the extracellular matrix and causes rounding and detachment of spread cells. This peptide is biotinylated through 6-aminohexanoate (LC) as a spacer.
Sequence:Biotin-LC-GRGDS
MW:830 Da
% peak area by HPLC:95
Storage condition:-20° C