The Wizard™ Classic line of random sparse matrix screens is designed to increase the probability of producing crystals during the coarse screening phase when crystallizing biological macromolecules (proteins, nucleic acids, peptides, and combinations thereof).
The Wizard™ Classic reagents are proven to be a highly effective starting point in the screening of biological macromolecules
Choose from Wizard™ Classic 1, 2, 3, or 4 non-overlapping formulations in matrix blocks or tubes. The 96-well block plate formulations are 1.7mL each in a 96 deep-well matrix block plate. Tube sets are 10mL volumes per tube. All formulations are prepared with ASTM type 1 water and high-purity chemicals, 0.2µm filtered in sterile packaging.
The Wizard Classic formulations include a large range of crystallants, buffers, and salts covering a broad range of crystallization space at pH levels from pH 4.5 to pH 10.5.