Order Entry
United States
ContactUsLinkComponent
QuickExtract™ RNA Extraction Solution, RT-PCR-ready RNA, Biosearch Technologies
QuickExtract™ RNA Extraction Solution, RT-PCR-ready RNA, Biosearch Technologies
Catalog # 76081-774
Supplier:  Lucigen
CAS Number:  
undefined
QuickExtract™ RNA Extraction Solution, RT-PCR-ready RNA, Biosearch Technologies
Catalog # 76081-774
Supplier:  Lucigen
Supplier Number:  QER090150
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
  • Method / Format:
    Extraction for direct PCR
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Sample type:
    Cultured cells
  • Target molecule:
    RNA
  • Description:
    QuickExtract™ RNA Extraction Kit
  • Size:
    50 mL
  • Environmentally Preferable:
  • Description:
    QuickExtract™ RNA Extraction Kit
  • Cat. No.:
    76081-774

Specifications

About this item

Rapid, simple RNA extraction (no purification) from mammalian cells, ready for RT-PCR applications

  • Fast: Extract end-point and real-time RT-PCR ready RNA in minutes from low to high numbers of samples
  • Compatible: Extracted RNA is compatible with both real-time and endpoint RT-PCR assays
  • Automation-friendly: Simple extraction protocol makes incorporation into an automated workflow easy
  • Flexible: When necessary, a DNase I digestion step can be added for sensitive downstream applications
  • Safe: No toxic reagents used

The QuickExtract™ RNA Extraction Kit is a fast, simple way of preparing RNA for RT-PCR (both end-point and real-time). The single-tube system requires only vortex mixing to lyse the cells, and prepare the RNA for cDNA synthesis. The result is easy processing of one to hundreds of samples in minutes, with no sample loss or toxic organic solvents. The QuickExtract RNA Extraction Solution works with cultured adherent and suspension cells including buccal cells, and has been tested on human, mouse, rat, E. coli, and S. aureus cell cultures. It is not suitable for tissue samples or plant samples. An optional DNase I treatment may improve certain downstream applications.