About this item
InVivoMab anti-human CD54 (ICAM-1), Clone: R6-5-D6, Isotype: mouse IgG2a, Applications: in vitro T cell stimulation/activation, Immunofluorescence
- Recommended Dilution Buffer: IP0065
Type: Primary
Antigen: CD54 (ICAM-1)
Clonality: Monoclonal
Clone: R6-5-D6
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 76555-980
- 76554-570
- 76556-300
- 76555-734
- 76557-340
- Antigen Symbol:
- ICAM-1
- CD54
- ICAM-1
- ICAM-1
- ICAM-1
- Antibody Type:
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- R6-5-D6
- R6-5-D6
- R6-5-D6
- R6-5-D6
- R6-5-D6
- Reactivity:
- Human
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG2a
- IgG2a
- IgG2a
- IgG2a
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Yes