ContactUsLinkComponent
Gas-Chex® EO Indicator Tape, Propper Manufacturing
Gas-Chex® EO Indicator Tape, Propper Manufacturing
Catalog # 76022-494
Supplier:  Avantor
undefined
Gas-Chex® EO Indicator Tape, Propper Manufacturing
Catalog # 76022-494
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Width:
    ¾"
  • Description:
    Gas-Chex® EO Indicator Tape
  • Length:
    60 yds.
  • Environmentally Preferable:
  • Cat. No.:
    76022-494
Specifications
About this item
Gas-Chex® indicator tape provides a means of identifying sterilized packs while ensuring their security during ethylene oxide sterilization.
  • Distinguish between processed and unprocessed sterilization packages
  • Strong crepe-embossed, latex-free tape
  • Distinct red to green color change after exposure to EO sterilization cycle
  • For use in securing wrapped surgical trays before, during, and after sterilization
Gas-Chex® Ethylene Oxide Gas Indicator Tape pressure-sensitive sterilization tape is used to distinguish between processed and unprocessed items. Gas-Chex® EO Gas Indicator Tape shows the word "gas" is written in reactive ink, after exposure to a EO sterilization cycle, 'gas' changes from red to green. Green means go! Gas-Chex® Tape has strong pressure-sensitive adhesive. Adhesive specifically formulated for secure bonding with fabric, non-woven wraps, paper, metal and glass, yet provides easy removal without traces of residue.
Certifications: AAMI/ISO 11140-1,Type 1
Packaging: Packaged 48 rolls per case.