To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Adipophilin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Guinea pig
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Environmentally Preferable:
- Format:Supplied as whole antiserum with 0.09% NaN3.
- Storage Temperature:Store at 4 deg C.
- Shipping Temperature:*Blue Ice*
- Immunogen:Adipophilin antibody was raised in guinea pig using a synthetic peptide corresponding to residues 1-29 of the N-terminus of human and murine adipophilin as the immunogen.
- Cat. No.:101986-464
- Supplier no.:20RAP002
Specifications
About this item
Adipophilin / ADRP (a member of the PAT family) is a ubiquitous component of lipid droplets. It has been found in milk fat globule membranes and on the surface of lipid droplets in various cultured cell lines (see e.g. Heid et al.; for review see e.g. Targett-Adams et al.); inducible by etomoxir. Enhanced expression of Adipophilin / ADRP is a useful marker for pathologies characterized by increased lipid droplet accumulation. Such diseases include atheroma, steatosis, obesity and certain cases of liposarcoma. It also seems to be a potent marker for atherosclerosis.
IHC-F: 1:100, IHC-P: 1:100, WB: 1:2,000
Type: Primary
Antigen: Adipophilin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Guinea Pig
Isotype:
Reactivity: Human