To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Acidic hair keratin K40
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Guinea pig
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Size:100 µl
- Environmentally Preferable:
- Format:Supplied in liquid form. Contains 0.09% sodium azide
- Storage Temperature:Store at 2-8 deg C for short term storage. Aliquot and store at -20 deg C for long term storage. Avoid repeated freeze/thaw cycles.
- Shipping Temperature:*Blue Ice*
- Immunogen:acidic hair keratin K40 antibody was raised in Guinea Pig using synthetic peptide of human type I (acidic) hair (trichocytic) keratin K40 coupled to KLH as the immunogen.
- Cat. No.:101986-416
- Supplier no.:20R2645
Keratin 40 is a member of the type I (acidic) keratin family, which belongs to the superfamily of intermediate filament (IF) proteins. Keratins are heteropolymeric structural proteins which form the intermediate filament. These filaments, along with actin microfilaments and microtubules, compose the cytoskeleton of epithelial cells. The type I keratin genes are clustered in a region of chromosome 17q12-q21.
IHC: 1:100
Type: Primary
Antigen: Acidic hair keratin K40
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Guinea Pig
Isotype:
Reactivity: Human