To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Plakophilin 3
- Clonality:Monoclonal
- Clone:PKP3-270.6.2
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:5 ml
- Environmentally Preferable:
- Format:Hybridoma culture supernatant.
- Storage Temperature:Store at 4 deg C.
- Shipping Temperature:*Blue Ice*
- Immunogen:Plakophilin 3 antibody was raised in mouse using recombinant protein (E.Coli) of human plakophilin 3 as the immunogen.
- Cat. No.:101983-796
- Supplier no.:10RP130C
Specifications
About this item
Plakophilin-3 is a protein that in humans is encoded by the PKP3 gene. This gene encodes a member of the arm-repeat (armadillo) and plakophilin gene families. Plakophilin proteins contain numerous armadillo repeats, localize to cell desmosomes and nuclei, and participate in linking cadherins to intermediate filaments in the cytoskeleton.
Optimal conditions to be determined by end user
Type: Primary
Antigen: Plakophilin 3
Clonality: Monoclonal
Clone: PKP3-270.6.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human