To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Keratin K75
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Guinea pig
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Environmentally Preferable:
- Format:Supplied in liquid form. Contains 0.09% sodium azide
- Storage Temperature:Store at 2-8 deg C for short term storage. Aliquot and store at -20 deg C for long term storage. Avoid repeated freeze/thaw cycles.
- Shipping Temperature:*Blue Ice*
- Immunogen:Keratin K75 antibody was raised in Guinea Pig using synthetic peptide of human type II keratin K75 coupled to KLH as the immunogen.
- Cat. No.:101986-420
- Supplier no.:20R2647
Specifications
About this item
Keratin 75 is a keratin protein. Keratin refers to a family of fibrous structural proteins. Keratin is the key structural material making up the outer layer of human skin. It is also the key structural component of hair and nails. Keratin monomers assemble into bundles to form intermediate filaments, which are tough and insoluble and form strong unmineralized tissues found in reptiles, birds, amphibians, and mammals. The only other biological matter known to approximate the toughness of keratinized tissue is chitin.
IHC: 1:100-200, WB: 1:2000
Type: Primary
Antigen: Keratin K75
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Guinea Pig
Isotype:
Reactivity: Human