To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:H+K+ ATPase
- Clonality:Monoclonal
- Clone:2G11
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Hog
- Western Blot:Yes
- Size:100 µl
- Environmentally Preferable:
- ChIP:Yes
- Format:PBS-diluted ascites containing 0.05% NaN3.
- Storage Temperature:Aliquot and store at -20 deg C. Avoid repeated freeze/thaw cycles.
- Concentration:1.0 mg/ml
- Shipping Temperature:*Blue Ice*
- Immunogen:H, K ATPase antibody was raised in mouse using a 34kDa core peptide purified from deglycosylated hog gastric microsomes as the immunogen.
- Cat. No.:101983-326
- Supplier no.:10RH100B
Specifications
About this item
The hydrogen/potassium ATPase, or gastric proton pump, belongs to a family of P-type cation-transporting ATPases. This family of ATPases shares a number of functional and structural similarities including the common feature of consisting of an alpha and beta subunit.
IF: 1:2,000, IHC-F: 1:200, IHC-P: 1:200 WB: 1:4,000
Type: Primary
Antigen: H+K+ ATPase
Clonality: Monoclonal
Clone: 2G11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Hog