To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:RPL37
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western Blot:Yes
- Size:50 µg
- Environmentally Preferable:
- Format:Lyophilized powder. Add 50 ul of distilled water. Final antibody concentration is 1 mg/ml in PBS buffer.
- Storage Temperature:Store at 4 deg C, following reconstitution, aliquot and store at -20 deg C. Avoid repeated freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:RPL37 antibody was raised in rabbit using the middle region of RPL37 as the immunogen
- Cat. No.:102016-578
- Supplier no.:70R10389
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L37E family of ribosomal proteins. It is located in the cytoplasm. The protein contains a C2C2-type zinc finger-like motif. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Optimal conditions to be determined by the end user
Type: Primary
Antigen: RPL37
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: