To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:hCGB
- Clonality:Monoclonal
- Clone:M48257
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:1 mg
- Environmentally Preferable:
- Format:PBS, pH 7.4, with 0.09% NaN3
- Storage Temperature:Store at 4 deg C for short term storage. Aliquot and store at -20 deg C for long term storage. Avoid repeated freeze/thaw cycles.
- Concentration:Batch dependent - please inquire should you have specific requirements
- Shipping Temperature:*Blue Ice*
- Immunogen:hCG Beta antibody was raised in Mouse using chorionic gonadotropin, B-subunit as the immunogen.
- Cat. No.:101977-218
- Supplier no.:10C25A
Specifications
About this item
Human chorionic gonadotropin (hCG) is a glycoprotein hormone produced in pregnancy that is made by the developing embryo after conception and later by the syncytiotrophoblast (part of the placenta). It is heterodimeric, with an alpha subunit identical to that of luteinizing hormone (LH), follicle-stimulating hormone (FSH), thyroid-stimulating hormone (TSH), and beta subunit that is unique to hCG.
Type: Primary
Antigen: hCGB
Clonality: Monoclonal
Clone: M48257
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human