To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Carbohydrate-responsive element-binding protein
- Antigen Symbol:MLXIPL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:400 ul
- Environmentally Preferable:
- Epitope:Amino Acid Range = 624-653
- Form:Liquid
- Gene ID:Q9NP71
- Antigen Synonyms:MLXIPL
- Modification:None
- Storage Buffer:Supplied in PBS with 0.09% (W/V) sodium azide.
- Molecular Weight:93 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.32 mg/mL
- Shipping Temperature:4 °C
- Immunogen:This MLXIPL antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 624-653 amino acids from the C-terminal region of human MLXIPL.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76012-006
Specifications
About this item
This gene encodes a basic helix-loop-helix leucine zipper transcription factor of the Myc/Max/Mad superfamily. This protein forms a heterodimeric complex and binds and activates, in a glucose-dependent manner, carbohydrate response element (ChoRE) motifs in the promoters of triglyceride synthesis genes. The gene is deleted in Williams-Beuren syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at chromosome 7q11.23.
For WB starting dilution is: 1:2000 For IHC-P starting dilution is: 1:10~50
Type: Primary
Antigen: MLXIPL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human, Mouse