To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Olfactory receptor 51S1
- Antigen Symbol:OR51S1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:400 ul
- Environmentally Preferable:
- Epitope:Amino Acid Range = 213-241
- Form:Liquid
- Gene ID:Q8NGJ8
- Antigen Synonyms:OR51S1
- Modification:None
- Storage Buffer:Supplied in PBS with 0.09% (W/V) sodium azide.
- Molecular Weight:35 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.45 mg/mL
- Shipping Temperature:4 °C
- Immunogen:This OR51S1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 213-241 amino acids from the C-terminal region of human OR51S1.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76011-784
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
For WB starting dilution is: 1:1000
Type: Primary
Antigen: OR51S1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human