Anti-ARHGAP10 Rabbit Polyclonal Antibody
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
GTPase activator for the small GTPases RhoA and Cdc42 by converting them to an inactive GDP-bound state. Essential for PTKB2 regulation of cytoskeletal organization via Rho family GTPases. Inhibits PAK2 proteolytic fragment PAK-2p34 kinase activity and changes its localization from the nucleus to the perinuclear region. Stabilizes PAK-2p34 thereby increasing stimulation of cell death (By similarity).
For WB starting dilution is: 1:1000 For IHC-P starting dilution is: 1:50~100 For FACS starting dilution is: 1:10~50
Type: Primary
Antigen: ARHGAP10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human
Specifications
- Size:400 ul
- Clonality:Polyclonal
- Modification:None
- Molecular Weight:89 kDa
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Epitope:Amino Acid Range = 237-263
- Form:Liquid
- Storage Buffer:Supplied in PBS with 0.09% (W/V) sodium azide.
- Host:Rabbit
- Concentration:0.5 mg/mL
- Cat. No.:76009-666
- Flow Cytometry:Yes
- Reactivity:Human
- Western Blot:Yes
- Shipping Temperature:4 °C
- Antibody Type:Primary
- Antigen Symbol:ARHGAP10
- Gene ID:A1A4S6
- Immunogen:This ARHGAP10 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 237-263 amino acids from the Central region of human ARHGAP10.
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen Synonyms:ARHGAP10
- Antigen Name:Rho GTPase-activating protein 10
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



