Gilian® Gilibrator-2 Diagnostic Kits, Sensidyne
Catalog # 87001-736
Supplier: Sensidyne
Small Business Enterprise
Specifications
- Description:Gilibrator-2 Deluxe Diagnostic Kit with Printer
- Environmentally Preferable:
- Cat. No.:87001-736
- Voltage:120 V
- Supplier no.:800844-4
Specifications
About this item
The Gilibrator-2 System provides convenient and highly automated way to check almost any commercially available air sampling pump for proper air flow function before deployment.
- Easy set-up and operation
- Field portable with battery or AC power
- Auto averaging of readings
- PC interface
Cell range is 20 cc/min to 6000 cc/min, 1 cc/min to 250 cc/min and 2 to 30 LPM.
Certifications: Traceable to NIST.
Delivery: Gilibrator-2 Diagnostic kits includes control base, three interchangeable wet cells for low, standard and high flow rates, DC charger/AC power supply, tubing and soap solution with dispenser, thermal printer, built-in diagnostic panel with rotameters and pressure gauge, in hard shell carry case.