Automated analyzer features advanced rheology technology.
- Results in 20 seconds
- 100μL sample
- No disposables
- Works directly off primary EDTA tubes
- Closed container sampling
- Fully automated
- Random access
- Continuous feed operation
- High throughput (up to 180 samples per hour)
- Internal barcode reader
- Internal printer
Utilizing a very small sample, this analyzer captures the impact of the most critical phase of the early-phase in the phenomenon of RBC sedimentation–the so-called Rouleaux formation. For the first time ever, ESR testing is fully automated and eliminates all of the traditional productivity roadblocks that have made ESR one of the most difficult and time-consuming tests in the laboratory.