ContactUsLinkComponent
Midi Plus™ Pipetting Controller, Sartorius
Midi Plus™ Pipetting Controller, Sartorius
Catalog # 75840-128
Supplier:  Sartorius
undefined
Midi Plus™ Pipetting Controller, Sartorius
Catalog # 75840-128
Supplier:  Sartorius
Supplier Number:  710931
Specifications
  • Description:
    Midi Plus™ Pipetting Controller
  • Environmentally Preferable:
  • Cat. No.:
    75840-128
  • Supplier no.:
    710931
Specifications
About this item
The lightweight, cordless Midi Plus pipetting controller enables users to aspirate from bottles and tubes without holding their arms and hands in an uncomfortable raised position.
  • Precise finger tip control of variable speed ranges
  • Hydrophobic autoclavable filter prevents over aspiration
  • Low-battery warning
Users can achieve fine-tune speed by applying varying finger pressure to the operating buttons.
The pipetting controller fits all commonly used 1 to 100 ml glass or plastic pipettes.
Tips
Product Information