2,4,6-Trinitrobenzenesulfonic acid (TNBS) 1% in methanol
Catalog # 89168-064
Supplier: G-Biosciences
CAS Number:
Specifications
- Size:10 ml
- Environmentally Preferable:
- Cat. No.:89168-064
- Supplier no.:BC86
Specifications
Specification Test Results
- Storage:Store at ‐20 °C
Specification Test Results
About this item
Synonyms: Picrylsulphonic acid solution, Picrylsulphonic acid, 2,4,6-Trinitrobenzenesulphonic acid
:
:293.17 g/mol
:GHS05
:GHS02
:GHS06
:GHS08
Storage Temperature:Freezer
CAS Number:2508-19-2
Formula:C₆H₃N₃O₉S
MDL Number:MFCD00064382