To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:CX3C-Chemokine Receptor 1 (HIV & Chemokine Receptor)
- Antigen Symbol:CX3CR1 / V28
- Clonality:Polyclonal
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Antigen Synonyms:CMKBRL1|V28
- Storage Buffer:Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage Temperature:Antibody can be stored at 4 °C for three months and at –20 °C for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Shipping Temperature:2-8°C
- Immunogen:Rabbit polyclonal CX3CR1 antibody was raised against a peptide corresponding to amino acids 2 to 21 of human CX3CR1 . The sequence differs from those of mouse and rat CX3CR1 by four amino acids
- Purification:Affinity chromatography Purified via peptide column.
- Cat. No.:10176-596
- Supplier no.:PK-AB718-2093
Rabbit Anti-Human/Mouse/Rat CX3C-Chemokine Receptor 1 (HIV & Chemokine Receptor)
CX3CR1 antibody can be used for detection of CX3CR1 by Western blot at 1:500 to 1:2000 dilution. An approximately 50 kDa band can be detected. Antibody can also be used for immunohistochemistry starting at 2 µg/mL. For immunofluorescence start at 10 µg/mL
Type: Primary
Antigen: CX3CR1 / V28
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat