Nanoject III™ Programmable Nanoliter Injector, Drummond
490019-810
: Drummond
- Description:Nanoject III Programmable Nanoliter Injector (100/240V)
- Environmentally Preferable:
- Cat. No.:490019-810
- Supplier no.:3-000-207
The Programmable Nanoject III Nanoliter Injector offers a number of features for easy setup and operation as well as more accurate injection volumes down to 0.6 nanoliters.
- User-Friendly Programmable Touchscreen Operation
- Positive Micropipet Attachments
- No O-Rings Required
- Easy Setup
- Hydraulic Technology for Accurate and Consistent Injection Volumes
The programmable touchscreen is user friendly, and the positive micropipet attachments do not require O-rings.
The injector's hydraulic technology provides accurate and consistent injection volumes.
: The kit contains control box, power source, injector head, injector head cable, 3.5" glass capillaries, 7" glass capillaries, backfilling needle, universal adapter and 4 interchangeable adapter plugs.