ContactUsLinkComponent
Difco™ Peptone Water, BD Diagnostics
Difco™ Peptone Water, BD Diagnostics
  90000-966
 :  
Difco™ Peptone Water, BD Diagnostics
  90000-966
 :  218071
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Difco™ Peptone Water
  • Size:
    500 g
  • Environmentally Preferable:
  • Cat. No.:
    90000-966
  • Supplier no.:
    218071
Peptone Water is used for cultivating nonfastidious organisms, for studying carbohydrate fermentation patterns, and for performing the indole test.
  • Peptone provides a source of carbon, nitrogen, vitamins, and minerals
  • Sodium chloride maintains osmotic equilibrium
  • Dehydrated appearance: Cream-white to light tan, free-flowing, homogeneous
The formulation of Peptone Water makes it useful for cultivating nonfastidious organisms. This nonselective medium has been used as a basal medium for biochemical tests such as carbohydrate fermentation patterns and production of indole.