Bacteria-3 Types Slide
Catalog # 470111-738
Supplier: United Scientific Supplies
Specifications
- Type:Monera
- Description:Bacteria- 3 Types
- Environmentally Preferable:
- Cat. No.:470111-738
- Supplier no.:100-06
Specifications
About this item
These slides help you compare and contrast bacteria morphology.
- Clearly Distinguishable Identifying Features
- Rigorous Quality Control Standards
- Slides Show Cocci, Bacillus, and Spirillum Bacterial Forms
They demonstrate cocci, bacillus, and spirillum bacterial forms in three separate smears.
The slides pass rigorous quality control standards.