Anti-SIGLEC6 Mouse Monoclonal Antibody (Alexa Fluor 700) [clone: 767329]
Catalog # 103238-700
Supplier: Novus Biologicals
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Sialic Acid Binding Ig-like Lectin 6
- Antigen Symbol:SIGLEC6
- Clonality:Monoclonal
- Clone:767329
- Conjugation:Alexa Fluor® 700
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 Tests
- Environmentally Preferable:
- Antigen Synonyms:CD33L|CD327|Sialic acid binding Ig-like lectin 6|Obesity-binding protein 1|OBBP1|CD33L2|CD33 antigen-like 1|CDW327|CD33L1|OB-BP1
- Storage Buffer:Supplied in a saline solution containing BSA and Sodium Azide.
- Storage Temperature:Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
- Immunogen:Chinese hamster ovary cell line CHO-derived recombinant human Siglec‑6/CD327 Gln27-Val331 Accession # NP_942142
- Cat. No.:103238-700
- Supplier no.:FAB2859N-100
Specifications
About this item
Type: Primary Antibodies
Antigen: SIGLEC6
Clonality: Monoclonal
Clone: 767329
Conjugation: Alexa Fluor 700
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human