Anti-SDPR Goat Polyclonal Antibody
103229-392
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Serum Deprivation Response
- Antigen Symbol:SDPR
- Clonality:Polyclonal
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:25 ug
- Environmentally Preferable:
- Antigen Synonyms:cavin-2|SDPR|CAVIN2|SDR|PS-p68|Cavin-2|Serum deprivation-response protein
- Storage Buffer:Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
- Storage Temperature:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months, -20 to -70 °C as supplied. 1 month, 2 to 8 °C sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
- Immunogen:E. coli-derived recombinant human SDPR Gly2-Val180 Accession # O95810
- Purification:Immunogen affinity purified
- Cat. No.:103229-392
- Supplier no.:AF5759-SP
Type: Primary Antibodies
Antigen: SDPR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse