Order Entry
United States
ContactUsLinkComponent
Acid Encapsulating & Neutralizing Mat Pad, PIG®
Acid Encapsulating & Neutralizing Mat Pad, PIG®
Catalog # 10147-158
Supplier:  New Pig
undefined
Acid Encapsulating & Neutralizing Mat Pad, PIG®
Catalog # 10147-158
Supplier:  New Pig
Supplier Number:  MAT304

Specifications

  • Description:
    PIG Acid Encapsulating & Neutralizing Mat Pad
  • Dimensions:
    11 W x 12"L (27.9 W x 30.4 L cm)
  • Environmentally Preferable:
  • Cat. No.:
    10147-158
  • Absorbency:
    3.2 oz./pad
  • Supplier no.:
    MAT304

Specifications

About this item

Encapsulate and neutralize the most common acid spills.

  • Great for battery charging, transport and storage areas
  • Identifies the best time to handle with the least amount of risk
  • Gelling agents solidify acids during neutralization

Encapsulate and neutralize the most common acid spills; great for battery charging, transport and storage areas. Color changes as liquids are taken in, then returns to normal when neutralization is complete; helps identify the best time to handle with the least amount of risk. Instantly absorbs up to 98% concentrations of sulfuric acid. Gelling agents solidify acids during neutralization and prevents them from leaking out of the mat.