To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Core-binding factor, runt Domain, alpha subunit 2; translocated to, 2
- Antigen Symbol:CBFA2T2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rabbit,Horse,Human,Bovine,Guinea pig,Dog,Rat,Mouse
- Western Blot:Yes
- Size:50 μg
- Environmentally Preferable:
- Gene ID:12396
- Antigen Synonyms:MTG8-like protein|MTG8-related protein 1|EHT|p85|CBFA2T2|ETO homologous on chromosome 20|MTGR1|Protein CBFA2T2|Myeloid translocation-related protein 1
- Storage Buffer:PBS buffer with 2% sucrose.
- Storage Temperature:For longer periods of storage, store at -20C. Avoid repeat freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:The immunogen for anti-CBFA2T2H antibody: synthetic peptide directed towards the middle region of mouse CBFA2T2H
- Cat. No.:10163-202
- Supplier no.:GTX44661
Rabbit polyclonal antibody to CBFA2T2
Type: Primary
Antigen: Cbfa2t2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Dog, Rat, Guinea Pig, Horse, Rabbit, Bovine