Order Entry
United States
ContactUsLinkComponent
Round Bottom Drum Liner, PIG®, New Pig
Round Bottom Drum Liner, PIG®, New Pig
Catalog # 10860-616
Supplier:  New Pig
undefined
Round Bottom Drum Liner, PIG®, New Pig
Catalog # 10860-616
Supplier:  New Pig
Supplier Number:  DRM929

Specifications

  • Capacity:
    55 gal.
  • Color:
    Clear
  • Description:
    Round bottom drum liner
  • Material:
    Polyethylene (LDPE)
  • Thickness:
    10 mil.
  • Environmentally Preferable:
  • Cat. No.:
    10860-616
  • Supplier no.:
    DRM929

Specifications

About this item

Protective liner lets you reuse drums and save on cleaning costs.

  • Reduces the cost associated with cleaning, reconditioning and disposing of used drums
  • Made from FDA Compliant resins for containment of food and cosmetic products
  • Liner's contoured design is easy to insert and won't leave folds or pockets that will trap liquid
  • Saves time cleaning and eliminates residue that could be a source of liability
  • Tie-off liner extends past the edge of the container for easy closure and tying

Ideal for storing paints, inks, coatings and other hard-to-clean materials.

For use with 55 gal. plastic or steel drums.

Certifications: FDA Compliant.