Disposable Cuvettes, CVD Series, Medicom
Supplier: Ocean Optics Inc
Certificates
About this item
CVD Series plastic cuvettes are versatile, no-maintenance sample cells for spectroscopy applications.
- Each cuvette has a 1 cm (10 mm) pathlength but varies by filling volume and window dimension
- Optional cuvette caps are available in blue, green, orange and yellow
- No-maintenance plastic cuvettes eliminate clean-up time
- Fill volume options – cuvettes for standard and microvolume sampling
- UV-Vis and Vis-NIR coverage - options for 220-900 nm and 350-900 nm wavelengths
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.