ContactUsLinkComponent
Stainless Steel Tube Brushes/Surface Brush, Restek
Stainless Steel Tube Brushes/Surface Brush, Restek
Catalog # RK20112
Supplier:  Restek
undefined
Stainless Steel Tube Brushes/Surface Brush, Restek
Catalog # RK20112
Supplier:  Restek
Supplier Number:  20112
Specifications
  • Bristle material:
    Stainless steel
  • Description:
    Stainless Steel Tube Brushes/Surface Brush
  • Environmentally Preferable:
  • Cat. No.:
    RK20112
  • Supplier no.:
    20112
Specifications
About this item
Unlike brass brushes that can leave a metal residue, these stainless steel tube brushes (3/8, 3/16 and 1/4") work well for cleaning dirty collectors, injector ports, and detector ports.
  • Stainless steel fabrication
  • No metal residue
The surface brush can be used to remove residue that builds up on metal detector jets and electronic contacts.