To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Environmentally Preferable:
- Tag sequence:C-6 His tag
- Storage Conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Endotoxin Content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Protein Synonyms:Delta transcription factor|INO80S|YY1|Transcriptional repressor protein YY1|NF-E1|Yin and yang 1|INO80 complex subunit S
- Protein/Peptide Name:Yin and Yang 1
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular Weight:12.6 kD
- Sequence:Val221-Gly321
- Formulation:Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested Applications:Bioassay
- Cat. No.:75791-144
- Supplier no.:92-275
Transcriptional repressor protein YY1(YY1)contains 4 C2H2-type zinc fingers and belongs to the YY transcription factor family. Multifunctional transcription factor exhibits positive and negative control on a large number of cellular and viral genes by binding to sites overlapping the transcription start site. The effect on transcription regulation of the protein is depending upon the context in which it binds and diverse mechanisms of action include direct activation or repression, indirect activation or repression via cofactor recruitment, or activation or repression by disruption of binding sites or conformational DNA changes. Its activity is regulated by transcription factors and cytoplasmic proteins that have been shown to abrogate or completely inhibit YY1-mediated activation or repression.
- Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.