Order Entry
United States
ContactUsLinkComponent
Human Recombinant IL2 (from Cells)
Human Recombinant IL2 (from Cells)
Catalog # 75790-510
Supplier:  Prosci
CAS Number:  
Human Recombinant IL2 (from Cells)
Catalog # 75790-510
Supplier:  Prosci
Supplier Number:  91-937
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Environmentally Preferable:
  • Tag sequence:
    C-6 His tag
  • Storage Conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Biological Activity:
    ED50 is less than 0.1 ng/ml as determined by the dose-dependent stimulation of murine CTLL-2 cells.
    Specific Activity of 1.0 x 107 IU/ mg.
  • Protein Synonyms:
    Interleukin-2|Aldesleukin|TCGF|IL-2|IL2|T-Cell Growth Factor
  • Protein/Peptide Name:
    IL2
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    16.4 kD
  • Sequence:
    Ala21-Thr153
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested Applications:
    Bioassay
  • Cat. No.:
    75790-510
  • Supplier no.:
    91-937

Specifications

About this item

Interleukin-2(IL-2) is an interleukin, a type of cytokine signaling molecule in the immune system,belongs to the IL-2 family. It is a powerful immunoregulatory lymphokine produced by T-cells in response to antigenic or mitogenic stimulation. IL-2/IL-2R signaling is required for T-cell proliferation and other fundamental functions that are essential for the immune response. IL-2 stimulates growth and differentiation of B-cells, NK cells, lymphokine-activated killer cells, monocytes, macrophages and oligodendrocytes.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.