To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:fission 1 (mitochondrial outer membrane) homolog (S. cerevisiae)
- Antigen Symbol:FIS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 ug
- Environmentally Preferable:
- Form:Liquid
- Gene ID:Q9Y3D6
- Antigen Synonyms:FIS1
- Storage Buffer:Liquid. In PBS containing 10% glycerol and 0.02% sodium azide.
- Storage Temperature:Stable for at least 1 year after receipt when stored at –20 °C.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Recombinant human Fis1.
- Tested Applications:Immunocytochemistry
- Cat. No.:75793-234
- Supplier no.:36-323
Specifications
About this item
Fis1 promotes the fragmentation of the mitochondrial network and its perinuclear clustering. It can induce cytochrome c release from mitochondria to the cytosol, ultimately leading to apoptosis. It also mediates peroxisomal fission.
Immunocytochemistry: (methanol fixation; 1:200). Immunoprecipitation: (1:200). Western Blot: (1:500).
Type: Primary
Antigen: FIS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat