About this item
Human AB serum is the preferred choice for work involving tissue engineering, T-Cells, regenerative medicine and cell therapy applications
- collected from healthy male AB donors
- Lack antibodies against A and B blood type antigens
- Tested and found negative for all required viral markers via FDA-approved methods
- Recommended for therapeutic use.
This significant decrease in serum allows for large cost savings and a decrease in variability, without sacrificing cell growth or performance
The increased demand combined with the rising prices and limited, sometimes questionable supply of fetal bovine serum (FBS) has become a recognized problem amongst cell culture scientists. Although dependent on cell type, serum concentrations can be reduced by 50% to 90% when used in combination with Zap-SR without sacrificing performance.
Zap-SR is a well-defined supplement that contains no human or animal components.