ContactUsLinkComponent
Human Corneal Epithelial Cells, Lifeline Cell Technology
Human Corneal Epithelial Cells, Lifeline Cell Technology
Catalog # 102970-670
CAS Number:  
undefined
Human Corneal Epithelial Cells, Lifeline Cell Technology
Catalog # 102970-670
Supplier Number:  FC-0029
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Cell Format:
    Cryopreserved
  • Description:
    Human Corneal Epithelial Cells
  • Size:
    500,000 cells per vial
  • Environmentally Preferable:
  • Cat. No.:
    102970-670
  • Supplier no.:
    FC-0029
Specifications
About this item
Lifeline® Normal Human Corneal Epithelial Cells are cryopreserved as secondary cells.
  • Isolated from human corneal tissue
  • Expanded twice in culture vessels
  • Quality tested to ensure optimal morphology and growth
  • Can be grown serum-free
  • No exposure to antimicrobials or phenol red
Lifeline® Normal Human Corneal Epithelial Cells are cryopreserved as secondary cells, e.g., cells are isolated from human corneal tissue and expanded twice in culture vessels before being harvested for cryopreservation
Lifeline® normal Human Corneal Epithelial Cells can be grown serum-free in OcuLife™ Medium. Corneal Epithelial Cells are not exposed to antimicrobials or phenol red.
Human Corneal Epithelial Cells are quality tested in OcuLife™ Medium to ensure optimal morphology and growth over a period of at least three passages.