Specifications
- Sterility:Non sterile
- Disposable/Reusable:Reusable
- Latex/Latex-free:Latex-Free
- Description:Collin Vaginal Speculum
- Material:Stainless steel
- Environmentally Preferable:
- Tip configuration:41/4×11/4"
- Curvature:Straight
- Cat. No.:10197-946
- Supplier no.:90-3945
Specifications
About this item
Sklar's® Collin Vaginal Speculum is a gynecological instrument used for performing pelvic examinations, Pap Smears, and examining the cervix.
- Double blade duckbill speculum inserted with the blades closed
- Industry standard
Unlike many other vaginal specula, there is no handle with the Collin device and the blades open laterally/horizontally versus vertically.