To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:NLRP12
- Clonality:Monoclonal
- Clone:3D11A53
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Gene ID:91662
- Antigen Synonyms:NACHT|PYRIN-containing APAF1-like protein 7 (PYPAF7)|Regulated by nitric oxide (RNO)|Monarch-1|LRR and PYD domains-containing protein 12
- Storage Temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C.
- Concentration:0.50 mg/mL
- Shipping Temperature:Ambient RT
- Immunogen:Partial human NLRP12 recombinant protein (93-1061 a.a.) expressed in E. coli.
- Purification:The antibody was purified by affinity chromatography.
- Cat. No.:10020-042
- Supplier no.:675202
Specifications
About this item
Purified anti-NLRP12 [3D11A53]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Mouse; Apps: WB; Size: 100 μg
Each lot of this antibody is quality control tested by Western blotting. For Western blotting, the suggested use of this reagent is 0.5 - 2.5 μg per ml. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: NLRP12
Clonality: Monoclonal
Clone: 3D11A53
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;Mouse