About this item
PerCP/Cy5.5 anti-mouse CX3CR1 [SA011F11]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.06 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CX3CR1
Clonality: Monoclonal
Clone: SA011F11
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 10020-336
- 10020-338
- Antigen Symbol:
- CX3CR1
- CX3CR1
- Antigen Synonyms:
- CMKBRL1|V28|CCRL1|CMKDR1|Fractalkine receptor|GPR13|Chemokine (C-X3-C motif) receptor 1
- CMKBRL1|V28|CCRL1|CMKDR1|Fractalkine receptor|GPR13|Chemokine (C-X3-C motif) receptor 1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
- PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
- Clone:
- SA011F11
- SA011F11
- Reactivity:
- Mouse
- Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 13051
- 13051
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Immunogen:
- Mouse CX3CR1-transfected cells
- Mouse CX3CR1-transfected cells
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography and conjugated with PerCP/Cy5.5 under optimal conditions. The solution is free of unconjugated PerCP/Cy5.5 and unconjugated antibody.
- The antibody was purified by affinity chromatography and conjugated with PerCP/Cy5.5 under optimal conditions. The solution is free of unconjugated PerCP/Cy5.5 and unconjugated antibody.
- Concentration:
- 0.20 mg/mL
- 0.20 mg/mL
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT