Anti-ASIC3 Rabbit Polyclonal Antibody
89515-702
: Abgent
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Acid Sensing Ion Channel Subunit 3
- Antigen Symbol:ASIC3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Environmentally Preferable:
- Epitope:N-Terminal
- Gene ID:9311
- Antigen Synonyms:Testis sodium channel 1|Amiloride sensitive cation channel 3|hTNaC1|hTNaC 1|ACCN 3|ASICs3|Neuronal amiloride-sensitive cation channel 3|DRASIC|ACCN3|ASIC3|ASIC 3|hASIC 3|Amiloride-sensitive cation channel 3|Proton gated cation channel subunit|Modulatory subunit of ASIC 2a|Acid sensing ion channel 3|Neuronal amiloride sensitive cation channel 3|SLNAC 1|Modulatory subunit of ASIC2a|Amiloride sensitive cation channel 3 testis|Acid-sensing ion channel 3|ASIC3_HUMAN|TNaC 1|hASIC3|SLNAC1|TNaC1.
- Amino Acid Number:55 to 83
- Storage Buffer:PBS with 0.09% (W/V) sodium azide, pH7.4
- Molecular Weight:58.905 kDa
- Sequence:FHHQTALDERESHRC
- Storage Temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.5mg/ml
- Shipping Temperature:4 °C
- Immunogen:This ACCN3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 55-83 amino acids from the N-terminal region of human ACCN3.
- Purification:Protein A & Peptide affinity purification
- Cat. No.:89515-702
- Supplier no.:AP19124A
Western Blot: 1:1000
Type: Primary
Antigen: ASIC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human