Order Entry
United States
ContactUsLinkComponent
Anti-TRA-2-54 Mouse Monoclonal Antibody [clone: TRA-2-54/2J]
Anti-TRA-2-54 Mouse Monoclonal Antibody [clone: TRA-2-54/2J]
Catalog # 10760-466
Supplier:  Biolegend
Anti-TRA-2-54 Mouse Monoclonal Antibody [clone: TRA-2-54/2J]
Catalog # 10760-466
Supplier:  Biolegend
Supplier Number:  358802
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    TRA-2-54
  • Clonality:
    Monoclonal
  • Clone:
    TRA-2-54/2J
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Size:
    100 μg
  • Environmentally Preferable:
  • Form:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Gene ID:
    249
  • Antigen Synonyms:
    ALPL|Liver/Bone/Kidney Alkaline Phosphatase
  • Storage Temperature:
    The antibody solution should be stored undiluted between 2 °C and 8 °C.
  • Concentration:
    0.50 mg/mL
  • Shipping Temperature:
    Ambient RT
  • Immunogen:
    Human embryonal carcinoma cells
  • Purification:
    The antibody was purified by affinity chromatography.
  • Cat. No.:
    10760-466
  • Supplier no.:
    358802

Specifications

About this item

Purified anti-human TRA-2-54 [TRA-2-54/2J]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, IF, IHC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: TRA-2-54
Clonality: Monoclonal
Clone: TRA-2-54/2J
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human