ContactUsLinkComponent
Berlind-Auvard Weighted Vaginal Speculum, OR Grade, Sklar
Berlind-Auvard Weighted Vaginal Speculum, OR Grade, Sklar
Catalog # 10193-340
Supplier:  Sklar
undefined
Berlind-Auvard Weighted Vaginal Speculum, OR Grade, Sklar
Catalog # 10193-340
Supplier:  Sklar
Supplier Number:  90-3558
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Berlind-Auvard Weighted Vaginal Speculum
  • Material:
    Stainless steel
  • Environmentally Preferable:
  • Curvature:
    Straight
  • Cat. No.:
    10193-340
  • Supplier no.:
    90-3558
Specifications
About this item
Sklar's Berling-Auvard Weighted Vaginal Speculum is a single-end vaginal speculum commonly used in OB/GYN procedures, such as vaginal hysterectomy or dilation and curettage.
The weight holds the speculum in place and provides an open view for the surgeon to perform procedures. This speculum is similar to a standard Auvard speculum but has a unique blade size. This product has a right angled, long, narrow blade, which is 41/4×15/8", with a medium 2.5 pound weight.