About this item
Silicone Vaginal Dilator Set contains four dilators, that can be used prior to surgical procedures and examinations for sexual rehabilitation following treatment for cancer, or for treatment of pelvic floor disorders such as vaginismus.
The set includes three different sizes (small, medium, large) of four dilators (small-extra large). This product is the set of large sized dilators.
The small dilator has an overall length of 120mm and outside diameter graduated from 19.5 to 24-mm. The medium dilator has an overall length of 128-mm and an outside diameter graduated from 22.5 to 28-mm. The large dilator has an overall length of 138-mm and an outside diameter graduated from 25.5 to 32-mm. The XL dilator has an overall length of 150-mm and an outside diameter graduated from 30 to 38-mm.