Anti-PIRB Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 326414]
Catalog # 220017-500
Supplier: R&D Systems
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:paired Ig-like receptor B
- Antigen Symbol:PIRB
- Clonality:Monoclonal
- Clone:326414
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Mouse
- Size:100 Tests
- Environmentally Preferable:
- Form:Supplied in a saline solution containing BSA and Sodium Azide.
- Gene ID:18733
- Antigen Synonyms:Gp91|LIR-3|Lilrb3|PIR-B
- Amino Acid Number:Gly24 to Gly635
- Storage Temperature:Protect from light. Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
- Shipping Temperature:The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
- Immunogen:Mouse myeloma cell line NS0-derived recombinant mouse PIR-B Gly24-Gly635; Accession # AAH26937
- Purification:Protein A or G purified from hybridoma culture supernatant
- Cat. No.:220017-500
- Supplier no.:FAB2754P
Specifications
About this item
Type: Primary
Antigen: PIRB
Clonality: Monoclonal
Clone: 326414
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse